Learn More
Lidocaine N-ethyl bromide, 99+%, Thermo Scientific Chemicals
Blocker of voltage-activated sodium channels
$231.26 - $231.26
Chemical Identifiers
| CAS | 21306-56-9 |
|---|---|
| Molecular Formula | C16H27BrN2O |
| Molecular Weight (g/mol) | 343.309 |
| MDL Number | MFCD00083182 |
| InChI Key | DLHMKHREUTXMCH-UHFFFAOYSA-N |
| Synonym | lidocaine n-ethyl bromide, qx 314 bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, 2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, qx-314 bromide, qx-314.br, n-ethyllidocaine bromide |
| PubChem CID | 9884487 |
| IUPAC Name | [2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide |
| SMILES | CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAJ60678MC
|
Thermo Scientific Chemicals
J60678MC |
100 mg |
Each for $231.26
|
|
|||||
Description
Blocker of voltage-activated sodium channels
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 21306-56-9 | |
| 343.309 | |
| DLHMKHREUTXMCH-UHFFFAOYSA-N | |
| 9884487 | |
| CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-] |
| C16H27BrN2O | |
| MFCD00083182 | |
| lidocaine n-ethyl bromide, qx 314 bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, 2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, qx-314 bromide, qx-314.br, n-ethyllidocaine bromide | |
| [2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide |
Specifications
| 21306-56-9 | |
| C16H27BrN2O | |
| 100 mg | |
| DLHMKHREUTXMCH-UHFFFAOYSA-N | |
| [2-(2,6-dimethylanilino)-2-oxoethyl]-triethylazanium;bromide | |
| 9884487 | |
| ≥99% | |
| Lidocaine N-ethyl bromide |
| White | |
| MFCD00083182 | |
| lidocaine n-ethyl bromide, qx 314 bromide, lidocaine n-ethyl bromide quaternary salt, n-2,6-dimethylphenylcarbamoylmethyl triethylammonium bromide, 2,6-dimethylphenyl carbamoyl methyl triethylazanium bromide, d0k6yk, qx-314 bromide, qx-314.br, n-ethyllidocaine bromide | |
| CC[N+](CC)(CC)CC(=O)NC1=C(C=CC=C1C)C.[Br-] | |
| 343.309 | |
| 343.31 | |
| Crystalline Solid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only