missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lidocaine Impurity G Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Specifications
Specifications
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Linear Formula | C13H20N2 O.HCl |
| Synonym | 2-(Isopropylamino)-2′,6′-acetoxylidide Hydrochloride |
| Type | Certified Reference Material |
| Quantity | 50 mg |
| Formula Weight | 256.77 |
| Grade | Certified Reference Material |