missing translation for 'onlineSavingsMsg'
Learn More

Lidocaine hydrochloride Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. p-7187400
Change view
Click to view available options
Quantity:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019573 500 mg
111018080 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111019573 Supplier Merck Emd Millipore Supplier No. PHR1257500MG
Only null left

Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Featured Industries: Pharmaceutical (small molecule)

Specifications

CAS 6108-05-0
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C14H22N2 O · HCl · H2O
MDL Number MFCD00150329
Synonym Lidocaine hydrochloride monohydrate; 2-Diethylamino-N-(2,6-dimethylphenyl)acetamide hydrochloride monohydrate; Lignocaine hydrochloride monohydrate; Xylocaine hydrochloride monohydrate
Type Certified Reference Material
Quantity 500 mg
Formula Weight 288.81
Grade Certified Reference Material