missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Triethylammonium bicarbonate buffer, 1 M, MilliporeSigma™ Supelco™
$242.79 - $811.19
Chemical Identifiers
| MDL Number | MFCD00466938 |
|---|---|
| Synonym | Triethylammonium hydrogen carbonate buffer |
Chemical Identifiers
| MFCD00466938 | |
| Triethylammonium hydrogen carbonate buffer |
Specifications
| 15715-58-9 | |
| 8.4 to 8.6 (20°C in neat) | |
| 100 mL | |
| For HPLC | |
| LiChropur |
| 1.015-1.024 | |
| MFCD00466938 | |
| Triethylammonium hydrogen carbonate buffer | |
| 1 M |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany