missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Triethylammonium bicarbonate buffer, 1 M, MilliporeSigma™ Supelco™

Catalog No. 111013261
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013261 500 mL
111013260 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013261 Supplier Merck Emd Millipore Supplier No. 18597500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00466938
Synonym Triethylammonium hydrogen carbonate buffer

Specifications

Density 1.015-1.024
CAS 15715-58-9
pH 8.4 to 8.6 (20°C in neat)
Quantity 500 mL
Grade For HPLC
Concentration or Composition (by Analyte or Components) 1 M
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany