missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Tetramethylammonium bisulfate , ≥99.0% (T), MilliporeSigma™ Supelco™
$322.32 - $1259.00
Chemical Identifiers
| MDL Number | MFCD00036149 |
|---|---|
| Synonym | Tetramethylammonium hydrogen sulfate |
Identifiants chimiques
| MFCD00036149 | |
| Tetramethylammonium hydrogen sulfate |
Spécifications
| 80526-82-5 | |
| (CH3)4 N(HSO4) | |
| Tetramethylammonium hydrogen sulfate | |
| ≥99.0% (T) | |
| LiChropur |
| 10 g | |
| MFCD00036149 | |
| 171.22 | |
| Ion Pair Chromatography |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany