missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetramethylammonium bisulfate , ≥99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111015595
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015595 10 g
111015596 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111015595 Supplier Merck Emd Millipore Supplier No. 8772410GF

Chemical Identifiers

MDL Number MFCD00036149
Synonym Tetramethylammonium hydrogen sulfate

Specifications

CAS 80526-82-5
Quantity 10 g
Linear Formula (CH3)4 N(HSO4)
Formula Weight 171.22
Percent Purity ≥99.0% (T)
Grade Ion Pair Chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany