Learn More
LiChropur™ Tetraheptylammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™
$312.88 - $1270.28
Chemical Identifiers
| MFCD00011861 |
Specifications
| 87°C to 89°C; 89°C to 91°C | |
| [CH3(CH2)6]4 N(Br) | |
| 10 g | |
| ≥99.0% (AT) | |
| LiChropur |
| 4368-51-8 | |
| MFCD00011861 | |
| 490.69 | |
| For ion pair chromatography |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany