missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Tetraheptylammonium bromide , ≥99.0% (AT), MilliporeSigma™ Supelco™

Catalog No. 111014203
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014203 50 g
111014202 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014203 Supplier Merck Emd Millipore Supplier No. 8729650GF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00011861

Specifications

Melting Point 87°C to 89°C; 89°C to 91°C
CAS 4368-51-8
Linear Formula [CH3(CH2)6]4 N(Br)
Quantity 50 g
Formula Weight 490.69
Percent Purity ≥99.0% (AT)
Grade For ion pair chromatography
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany