missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium Phosphate Monobasic, MilliporeSigma™ Supelco™
Specifications
Specifications
| pH Range | 4 to 4.5 (20°C, 10% in H2O) |
| Purity or Quality Grade | 99% to 101% (T) |
| Quantity | 50 g |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany