missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium dodecyl sulfate,, For ion pair chromatography, ≥99.0%, MilliporeSigma™ Supelco™
Sodium dodecyl sulfate (SDS) is a detergent suitable to denature proteins. It is a long-chain aliphatic group which has negatively charged sulfate, hence making it amphipathic detergent in nature.
Supplier: Merck Emd Millipore 7172650GF
Specifications
| 204°C to 207°C | |
| 50 g | |
| MFCD00036175 | |
| Dodecyl sodium sulfate; Dodecyl sulfate sodium salt; Lauryl sulfate sodium salt; SDS; Sodium lauryl sulfate | |
| 288.38 | |
| Ion Pair Chromatography |
| 151-21-3 | |
| CH3(CH2)11 OSO3 Na | |
| 170°C | |
| Water: soluble | |
| ≥99.0% (GC); ≥99.0% | |
| LiChropur |
Chemical Identifiers
| MFCD00036175 | |
| Dodecyl sodium sulfate; Dodecyl sulfate sodium salt; Lauryl sulfate sodium salt; SDS; Sodium lauryl sulfate |
Safety and Handling
RTECSNumber : WT1050000
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany