missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium acetate, 99.0-101.0% (NT), MilliporeSigma™ Supelco™
Supplier: Merck Emd Millipore 79714250G
Specifications
| >300°C | |
| 127-09-3 | |
| 250 g | |
| MFCD00012459 | |
| Acetic acid sodium salt | |
| 82.03 | |
| HPLC | |
| LiChropur |
| 8.5 to 10.5 | |
| Solid | |
| CH3 COONa | |
| >250°C - Closed Cup | |
| Water: 10%, clear, colorless | |
| 99.0 to 101.0% (NT) | |
| 1112°F |
Chemical Identifiers
| MFCD00012459 | |
| Acetic acid sodium salt |
Safety and Handling
RTECSNumber : AJ4300010
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany