missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium acetate, 99.0-101.0% (NT), MilliporeSigma™ Supelco™

Catalog No. p-7187900
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012888 50 g
111012887 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111012888 Supplier Merck Emd Millipore Supplier No. 7971450G
Only null left

Chemical Identifiers

MDL Number MFCD00012459
Synonym Acetic acid sodium salt

Specifications

Melting Point >300°C
pH 8.5 to 10.5
Physical Form Solid
CAS 127-09-3
Quantity 50 g
Linear Formula CH3 COONa
Flash Point >250°C - Closed Cup
Solubility Information Water: 10%, clear, colorless
Formula Weight 82.03
Percent Purity 99.0 to 101.0% (NT)
Grade HPLC
Ignition Point 1112°F
Product Line LiChropur
Show More Show Less
RTECSNumber AJ4300010

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany