Learn More
LiChropur™ Sodium 1-pentanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
Sodium 1-pentanesulfonate monohydrate is an Ion pair reagent. It is a anionic surfactant which has opposite influence on aniline polymerization thermal effect. It also has highest and lowest enthalpy of polymerization values.
Supplier: Merck Emd Millipore 7695250GF
Specifications
| 207605-40-1 | |
| MFCD00149548 | |
| 1-Pentanesulfonic acid sodium salt | |
| ≥99.0% (T) | |
| LiChropur |
| CH3(CH2)4 SO3 Na · H2O | |
| 50 g | |
| 192.21 | |
| For ion pair chromatography |
Chemical Identifiers
| MFCD00149548 | |
| 1-Pentanesulfonic acid sodium salt |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany