missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-pentanesulfonate monohydrate , ≥99.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 207605-40-1 |
| Linear Formula | CH3(CH2)4 SO3 Na · H2O |
| Quantity | 5 g |
| Formula Weight | 192.21 |
| Percent Purity | ≥99.0% (T) |
| Grade | For ion pair chromatography |
| Product Line | LiChropur |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany