missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Sodium 1-heptanesulfonate monohydrate, 99.0% (T), MilliporeSigma™ Supelco™
Specifications
Specifications
| Description | Anionic |
| CAS | 207300-90-1 |
| Product Line | LiChropur |
| Linear Formula | CH3(CH2)5 CH2 SO3 Na · H2O |
| MDL Number | MFCD00149550 |
| Synonym | 1-Heptanesulfonic acid sodium salt monohydrate |
| Type | Organic Standard |
| Quantity | 10 g |
| Formula Weight | 220.26 |
| Percent Purity | ≥99% (T) |
| Show More |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany