missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Sodium 1-heptanesulfonate monohydrate, 99.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111013495
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013495 10 g
111013497 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111013495 Supplier Merck Emd Millipore Supplier No. 5183210GF

Specifications

Description Anionic
CAS 207300-90-1
Product Line LiChropur
Linear Formula CH3(CH2)5 CH2 SO3 Na · H2O
MDL Number MFCD00149550
Synonym 1-Heptanesulfonic acid sodium salt monohydrate
Type Organic Standard
Quantity 10 g
Formula Weight 220.26
Percent Purity ≥99% (T)
Grade Ion Pair Chromatography
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany