missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ (S)-(+)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7193494
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111018833 100 mg Bottomless Glass Bottle with Fused Cone
111018050 500 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018833 Supplier Merck Emd Millipore Supplier No. 65365100MGF
Only null left

For Chiral Derivatization

Chemical Identifiers

CAS 20445-33-4
Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00067105
InChI Key PAORVUMOXXAMPL-SECBINFHSA-N
Synonym (S)-(+)-MTPA-Cl; Mosher's acid chloride
IUPAC Name (2S)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Density 1.35 g/mL (at 25°C)
Boiling Point 213°C to 214°C
Packaging Bottomless Glass Bottle with Fused Cone
Linear Formula C6H5C(OCH3)(CF3)COCl
Refractive Index n20/D 1.469; n20/D 1.469
Quantity 100 mg
Formula Weight 252.62
Percent Purity ≥99.0% (AT)
Grade For chiral derivatization

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany