missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Phosphoric Acid, MilliporeSigma™ Supelco™

For HPLC

$595.63 - $595.63

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111022598
View Documents
Merck Emd Millipore
49685500ML
500 mL
Each for $595.63
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7664-38-2
97.99
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
H3O4P
MFCD00011340
Orthophosphoric Acid
OP(O)(O)=O

Specifications

∼40°C (literature)
158°C (literature)
2.2 mmHg (20°C)
H3O4P
MFCD00011340
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
98
HPLC
7664-38-2
1.685 g/mL at 25°C (literature)
500 mL
H3PO4
Orthophosphoric Acid
OP(O)(O)=O
97.99
85%
Videos
Safety and Handling

Safety and Handling

H290 - H314

RTECSNumber : TB6300000

SDS
Documents

Documents

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany