missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Phosphoric Acid, MilliporeSigma™ Supelco™

Catalog No. 111022598
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022598 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022598 Supplier Merck Emd Millipore Supplier No. 49685500ML

missing translation for 'validMainframeMsg'

For HPLC

Chemical Identifiers

CAS 7664-38-2
Molecular Formula H3O4P
Molecular Weight (g/mol) 97.99
MDL Number MFCD00011340
InChI Key NBIIXXVUZAFLBC-UHFFFAOYSA-N
Synonym Orthophosphoric Acid
IUPAC Name phosphoric acid
SMILES OP(O)(O)=O

Specifications

Melting Point ∼40°C (literature)
Boiling Point 158°C (literature)
Density 1.685 g/mL at 25°C (literature)
Vapor Pressure 2.2 mmHg (20°C)
Quantity 500 mL
Linear Formula H3PO4
Formula Weight 98
Percent Purity 85%
Grade HPLC
Hazard Category H290 - H314
RTECSNumber TB6300000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany