missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide, MilliporeSigma™ Supelco™

Catalog No. 111022774
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022774 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022774 Supplier Merck Emd Millipore Supplier No. 7762610ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide is a silylating reagent which replaces the active hydrogen with tert-Butyldimethylsilyl group. The tert-Butyldimethylsilyl derivatives are found to be more resistant to hydrolysis and stable compared to trimethylsilyl (TMS) derivatives.

  • Suitable for GC-MS analysis since it produces mass spectra, which can be easily interpreted
  • It is widely used for the silylation of different alcohols, thiols, phenols, carboxylic acids, amines, and amides

Chemical Identifiers

CAS 77377-52-7
Molecular Formula C9H18F3NOSi
Molecular Weight (g/mol) 241.33
MDL Number MFCD00009671
InChI Key NEJIKZHUEMMAEF-UHFFFAOYSA-N
Synonym MTBSTFA
IUPAC Name N-(butyldimethylsilyl)-2,2,2-trifluoro-N-methylacetamide
SMILES CCCC[Si](C)(C)N(C)C(=O)C(F)(F)F

Specifications

Density 1.036 g/mL (at 25°C (literature))
Flash Point 45°C (Closed Cup)
Linear Formula CF3CON(CH3)Si(CH3)2 C(CH3)3
UN Number UN 1993 C 3 / PGIII
Boiling Point 172°C to 175°C (literature)
Quantity 10 mL
Formula Weight 241.33
Percent Purity ≥99% (GC)
Grade GC Derivatization
Hazard Category H226 - H315 - H319 - H335
Recommended Storage 2°C to 8°C

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany