missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, ≥98.5-99%, MilliporeSigma™ Supelco™
$42.40 - $1150.01
Chemical Identifiers
| MDL Number | MFCD00008269 |
|---|---|
| Synonym | Silylating mixture V; BSTFA + TMCS |
Chemical Identifiers
| MFCD00008269 | |
| Silylating mixture V; BSTFA + TMCS |
Specifications
| 25561-30-2 | |
| 45°C to 50°C (14 mmHg) | |
| Glass Bottle | |
| MFCD00008269 | |
| Silylating mixture V; BSTFA + TMCS | |
| ≥98.5% (excluding chlorotrimethylsilane, GC); 99% (excluding TMCS) | |
| LiChropur |
| 0.97 g/mL | |
| 6°C | |
| CF3 C[=NSi(CH3)3 ]OSi(CH3)3 | |
| 1 mL | |
| 257.40 | |
| For GC derivatization |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany