missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N,O-Bis(trimethylsilyl)trifluoroacetamide with trimethylchlorosilane, ≥98.5-99%, MilliporeSigma™ Supelco™

Catalog No. 111020670
Change view
Click to view available options
Quantity:
1 mL
5 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020670 1 mL
111020237 5 mL
111018072 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111020670 Supplier Merck Emd Millipore Supplier No. 152381ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00008269
Synonym Silylating mixture V; BSTFA + TMCS

Specifications

CAS 25561-30-2
Density 0.97 g/mL
Boiling Point 45°C to 50°C (14 mmHg)
Flash Point 6°C
Packaging Glass Bottle
Linear Formula CF3 C[=NSi(CH3)3 ]OSi(CH3)3
Quantity 1 mL
Molecular Weight (g/mol) 257.40
Formula Weight 257.40
Percent Purity ≥98.5% (excluding chlorotrimethylsilane, GC); 99% (excluding TMCS)
Grade For GC derivatization
Physical Form Liquid
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany