Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 98.5%, MilliporeSigma™ Supelco™
N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a silylating reagent.
Supplier:  Merck Emd Millipore 6947910X1ML
 View more versions of this product
Catalog No. 111018706
Only null left
Explore available promotions
Description
Description
Specifications
Specifications
24589-78-4
8.8 mmHg (27°C)
1.075 g/mL (at 25°C)
CF3 CON(CH3)Si(CH3)3
130°C to 132°C
Organic Standard
199.25
GC Derivatization
LiChropur
>1 (vs. air)
26°C - closed cup
MFCD00000411
N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA
10 x 1 mL
≥98.5%
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Promotions
Promotions
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany