missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ N-Methyl-N-(trimethylsilyl)trifluoroacetamide, 98.5%, MilliporeSigma™ Supelco™
N-Methyl-N-(trimethylsilyl)trifluoroacetamide (MSTFA) is a silylating reagent.
Supplier: Merck Emd Millipore 6947925ML
Specifications
| 24589-78-4 | |
| 8.8 mmHg (27°C) | |
| 1.075 g/mL (at 25°C) | |
| CF3 CON(CH3)Si(CH3)3 | |
| 130°C to 132°C | |
| Organic Standard | |
| 199.25 | |
| GC Derivatization |
| LiChropur | |
| >1 (vs. air) | |
| 26°C - closed cup | |
| MFCD00000411 | |
| N-Trimethylsilyl-N-methyl trifluoroacetamide; MSTFA | |
| 25 mL | |
| ≥98.5% |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany