missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ N-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide, For chiral derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. 111015964
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015964 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015964 Supplier Merck Emd Millipore Supplier No. 7147850MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Nα-(2,4-Dinitro-5-fluorophenyl)-L-alaninamide (FDAA) is a chiral derivatizing agent (CDA), has high enantioselectivity but low sensitivity as compared to other CDAs. It is generally used to assign the stereochemistry of amino acids in trace amounts.

Chemical Identifiers

MDL Number MFCD00042049
Synonym FDAA; Marfey′s reagent

Specifications

CAS 95713-52-3
Linear Formula C9H9 FN4O5
Quantity 50 mg
Formula Weight 272.19
Percent Purity ≥99.0% (sum of enantiomers, TLC); ≥99.0%
Grade For chiral derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany