missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Methoxyamine Hydrochloride, MilliporeSigma™ Supelco™
Methoxyamine hydrochloride is a commonly used reagent for the generation of oxime derivatives of aldehyde- or ketone-group containing compounds. It is typically employed as an oximation reagent prior to silylation derivatizations.
$88.17 - $296.02
Chemical Identifiers
| CAS | 593-56-6 |
|---|---|
| Molecular Formula | CH6ClNO |
| Molecular Weight (g/mol) | 83.52 |
| MDL Number | MFCD00012951 |
| InChI Key | XNXVOSBNFZWHBV-UHFFFAOYSA-N |
| Synonym | O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride |
| IUPAC Name | hydrogen O-methylhydroxylamine chloride |
| SMILES | [H+].[Cl-].CON |
Chemical Identifiers
| 593-56-6 | |
| 83.52 | |
| XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
| hydrogen O-methylhydroxylamine chloride |
| CH6ClNO | |
| MFCD00012951 | |
| O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride | |
| [H+].[Cl-].CON |
Specifications
| 593-56-6 | |
| CH6ClNO | |
| MFCD00012951 | |
| UN3261 - Class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all | |
| XNXVOSBNFZWHBV-UHFFFAOYSA-N | |
| hydrogen O-methylhydroxylamine chloride | |
| 83.52 | |
| For GC Derivatization |
| 151°C to 154°C (literature) | |
| CH3ONH2 · HCl | |
| 1 g | |
| O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride | |
| [H+].[Cl-].CON | |
| 83.52 | |
| 97.5% to 102.5% (AT) |
Safety and Handling
H290 - H302 + H312 - H315 - H317 - H319 - H351 - H372 - H410
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany