missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Methoxyamine Hydrochloride, MilliporeSigma™ Supelco™

Catalog No. 111022798
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022798 5 g
111022797 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111022798 Supplier Merck Emd Millipore Supplier No. 898035G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Methoxyamine hydrochloride is a commonly used reagent for the generation of oxime derivatives of aldehyde- or ketone-group containing compounds. It is typically employed as an oximation reagent prior to silylation derivatizations.

Chemical Identifiers

Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.52
MDL Number MFCD00012951
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym O-Methylhydroxylamine hydrochloride; Methoxylamine hydrochloride
IUPAC Name hydrogen O-methylhydroxylamine chloride
SMILES [H+].[Cl-].CON

Specifications

Melting Point 151°C to 154°C (literature)
Linear Formula CH3ONH2 · HCl
Quantity 5 g
UN Number UN3261 - Class 8 - PG 2 - EHS - Corrosive solid, acidic, organic, n.o.s., HI: all
Formula Weight 83.52
Percent Purity 97.5% to 102.5% (AT)
Grade For GC Derivatization
Hazard Category H290 - H302 + H312 - H315 - H317 - H319 - H351 - H372 - H410

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany