missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ ™-(-)-α-Methoxy-α-(trifluoromethyl)phenylacetyl chloride, 99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7193516
Change view
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Bottomless Glass Bottle with Fused Cone
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111018938 100 mg Bottomless Glass Bottle with Fused Cone
111018083 500 mg Bottomless Glass Bottle with Fused Cone
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111018938 Supplier Merck Emd Millipore Supplier No. 65363100MG
Only null left

For Chiral Derivatization

™-(-)-a-Methoxy-a-(trifluoromethyl)phenylacetyl chloride (Mosher's acid chloride) is generally used as a chiral derivatizing agent. It is easily available in enantiomerically pure form. It can form diastereomers with both alcohols and amines.

Chemical Identifiers

Molecular Formula C10H8ClF3O2
Molecular Weight (g/mol) 252.62
MDL Number MFCD00044400
InChI Key PAORVUMOXXAMPL-VIFPVBQESA-N
Synonym (R)-(-)-MTPA-Cl; Mosher's acid chloride
IUPAC Name (2R)-3,3,3-trifluoro-2-methoxy-2-phenylpropanoyl chloride
SMILES CO[C@](C(Cl)=O)(C1=CC=CC=C1)C(F)(F)F

Specifications

Density 1.35 g/mL (at 25°C)
Boiling Point 213°C to 214°C
Packaging Bottomless Glass Bottle with Fused Cone
Refractive Index n20/D 1.469; n20/D 1.47
Linear Formula C6H5C(OCH3)(CF3)COCl
Quantity 100 mg
Formula Weight 252.62
Percent Purity ≥99.0% (AT)
Grade For chiral derivatization
Product Line LiChropur

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany