missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Methanesulfonic Acid, For HPLC, 99.5% (T), MilliporeSigma™ Supelco™

Catalog No. p-7193376
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Packaging:
Clear Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
111020290 1 mL Clear Glass Bottle
111018808 10 x 1 mL Clear Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111020290 Supplier Merck Emd Millipore Supplier No. 595101ML
Only null left

Chemical Identifiers

CAS 75-75-2
Molecular Formula CH4O3S
Molecular Weight (g/mol) 96.10
MDL Number MFCD00007518
InChI Key AFVFQIVMOAPDHO-UHFFFAOYSA-N
IUPAC Name methanesulfonic acid
SMILES CS(O)(=O)=O

Specifications

Boiling Point 167°C (10 mmHg)
Melting Point 17°C to 19°C
Color APHA: ≤25
Quantity 1 mL
Linear Formula CH3SO3H
Formula Weight 96.11
Percent Purity ≥99.5% (T)
Grade HPLC
Packaging Clear Glass Bottle
Refractive Index n20/D 1.428-1.430; n20/D 1.429
Vapor Pressure 1 mmHg (20°C)
Density 1.481 g/mL (at 25°C); Vapor Density: 3.3 (vs. air)
Show More Show Less
RTECSNumber PB1140000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany