missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Hydrindantin dihydrate for Stein-Moore-Chromatography, MilliporeSigma™ Supelco™

Catalog No. p-7187465
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014803 10 g
111014802 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 111014803 Supplier Merck Emd Millipore Supplier No. 5394010G
Only null left

Specifications

CAS 5950-69-6
Product Line LiChropur
Linear Formula C18H14O8
MDL Number MFCD00149242
Melting Point 240°C
For Use With (Equipment) Stein-Moore-Chromatograph
Synonym 2,2′,3,3,3′,3′-Hexahydroxy-2,2′-biindan-1,1′-dione
Type Organic Standard
Quantity 10 g
Formula Weight 358.30

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany