missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Hydrindantin dihydrate for Stein-Moore-Chromatography, MilliporeSigma™ Supelco™
Specifications
Specifications
| CAS | 5950-69-6 |
| Product Line | LiChropur |
| Linear Formula | C18H14O8 |
| MDL Number | MFCD00149242 |
| Melting Point | 240°C |
| For Use With (Equipment) | Stein-Moore-Chromatograph |
| Synonym | 2,2′,3,3,3′,3′-Hexahydroxy-2,2′-biindan-1,1′-dione |
| Type | Organic Standard |
| Quantity | 50 g |
| Formula Weight | 358.30 |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany