missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Heptafluorobutyric anhydride, For GC derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Supplier:  Merck Emd Millipore 7725350ML

 View more versions of this product

Catalog No. 111018000

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

-43°C
1.674 g/mL at 20°C; 1.674 g/mL (at 20°C)
Ampule
MFCD00000432
HFAA; HFBA; Perfluorobutyric anhydride
≥99.0% (GC); ≥99.0%
LiChropur
336-59-4
108°C to 110°C
(CF3 CF2 CF2 CO)2 O
50 mL
410.06
For GC derivatization

Chemical Identifiers

MFCD00000432
HFAA; HFBA; Perfluorobutyric anhydride
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany