missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Heptafluorobutyric anhydride, For GC derivatization, ≥99.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187742
Change view
Click to view available options
Quantity:
10 mL
50 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018645 10 x 1 mL
111018667 10 mL
111018000 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111018645 Supplier Merck Emd Millipore Supplier No. 7725310X1ML
Only null left

Chemical Identifiers

MDL Number MFCD00000432
Synonym HFAA; HFBA; Perfluorobutyric anhydride

Specifications

Melting Point -43°C
Density 1.674 g/mL at 20°C; 1.674 g/mL (at 20°C)
CAS 336-59-4
Boiling Point 108°C to 110°C
Packaging Ampule
Linear Formula (CF3 CF2 CF2 CO)2 O
Quantity 10 x 1 mL
Formula Weight 410.06
Percent Purity ≥99.0% (GC); ≥99.0%
Grade For GC derivatization
Product Line LiChropur
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany