missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ Ammonium phosphate dibasic, 99.0-101.0% (T), MilliporeSigma™ Supelco™
$86.58 - $86.58
Chemical Identifiers
| MDL Number | MFCD00010891 |
|---|---|
| Synonym | Ammonium hydrogenphosphate; Diammonium hydrogenphosphate; di-Ammonium hydrogenphosphate (sec) |
Chemical Identifiers
| MFCD00010891 | |
| Ammonium hydrogenphosphate; Diammonium hydrogenphosphate; di-Ammonium hydrogenphosphate (sec) |
Specifications
| 155°C | |
| 250 g | |
| MFCD00010891 | |
| Water: 50 mg/mL, clear, colorless | |
| 99.0 to 101.0% (T) | |
| LiChropur |
| 7783-28-0 | |
| (NH4)2 HPO4 | |
| Ammonium hydrogenphosphate; Diammonium hydrogenphosphate; di-Ammonium hydrogenphosphate (sec) | |
| 132.06 | |
| HPLC |
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany