missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ Ammonium phosphate dibasic, 99.0-101.0% (T), MilliporeSigma™ Supelco™

Catalog No. 111015520
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015520 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015520 Supplier Merck Emd Millipore Supplier No. 79762250G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

MDL Number MFCD00010891
Synonym Ammonium hydrogenphosphate; Diammonium hydrogenphosphate; di-Ammonium hydrogenphosphate (sec)

Specifications

Melting Point 155°C
CAS 7783-28-0
Physical Form Crystals
Quantity 250 g
Linear Formula (NH4)2 HPO4
Solubility Information Water: 50 mg/mL, clear, colorless
Molecular Weight (g/mol) 132.06
Formula Weight 132.06
Percent Purity 99.0 to 101.0% (T)
Grade HPLC
Product Line LiChropur
Chemical Name or Material Ammonium Phosphate Dibasic
Show More Show Less

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany