missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LiChropur™ 1-(Trimethylsilyl)imidazole, ≥94.0%, MilliporeSigma™ Supelco™
Powerful silylating agent, particularly for alcohols; Synthesis of acyl imidazolides
Supplier: Merck Emd Millipore 39487425ML
Specifications
| 18156-74-6 | |
| 93°C to 94°C (14 mmHg) | |
| Ampule | |
| MFCD00005280 | |
| TSIM; N-Trimethylsilylimidazole; TMSI; TSIM | |
| ≥94.0% | |
| LiChropur |
| 0.956 g/mL (at 25°C) | |
| 66°C | |
| C6H12N2 Si | |
| 25 mL | |
| 140.26 | |
| For GC derivatization |
Chemical Identifiers
| MFCD00005280 | |
| TSIM; N-Trimethylsilylimidazole; TMSI; TSIM |
Safety and Handling
RTECSNumber : NI8700000
LiChropur is a trademark of Merck KGaA, Darmstadt, Germany