missing translation for 'onlineSavingsMsg'
Learn More

LiChropur™ 1-(Trimethylsilyl)imidazole, ≥94.0%, MilliporeSigma™ Supelco™

Catalog No. p-7187981
Change view
Click to view available options
Quantity:
5 mL
25 mL
10 x 1 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019375 5 mL
111018785 10 x 1 mL
111018454 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 111019375 Supplier Merck Emd Millipore Supplier No. 3948745ML
Only null left

Powerful silylating agent, particularly for alcohols; Synthesis of acyl imidazolides

Chemical Identifiers

MDL Number MFCD00005280
Synonym TSIM; N-Trimethylsilylimidazole; TMSI; TSIM

Specifications

Density 0.956 g/mL (at 25°C)
CAS 18156-74-6
Boiling Point 93°C to 94°C (14 mmHg)
Flash Point 66°C
Packaging Ampule
Linear Formula C6H12N2 Si
Quantity 5 mL
Formula Weight 140.26
Percent Purity ≥94.0%
Grade For GC derivatization
Product Line LiChropur
Show More Show Less
RTECSNumber NI8700000

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany