missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LHX2 Recombinant Rabbit Monoclonal Antibody (23GB6675)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA553093
Description
LHX2 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot
This gene encodes a protein belonging to a large protein family, members of which carry the LIM domain, a unique cysteine-rich zinc-binding domain. The encoded protein may function as a transcriptional regulator. The protein can recapitulate or rescue phenotypes in Drosophila caused by a related protein, suggesting conservation of function during evolution.Specifications
| LHX2 | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P36198, P50458, Q9Z0S2 | |
| Lhx2 | |
| A synthesized peptide derived from human LHX2 (1-56AA). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB6675 | |
| Unconjugated | |
| Lhx2 | |
| ap; apterous; hLhx2; Homeobox protein LH-2; LH2; Lh-2; LH2A; LHX2; LIM homeo box protein 2; LIM homeobox 2; LIM homeobox protein 2; LIM HOX gene 2; LIM/homeobox protein Lhx2; Lim2; MGC138390 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 16870, 296706, 9355 | |
| -20°C | |
| Liquid |