missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LGR6 Recombinant Rabbit Monoclonal Antibody (23GB6400)
GREENER_CHOICE

Catalog No. MA553038
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553038 100 μL
MA553039 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553038 Supplier Invitrogen™ Supplier No. MA553038
Only null left

Rabbit Recombinant Monoclonal Antibody

LGR6 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

LGR6 has been reported to be expressed in rat adrenal, brain, colon, heart, kidney, ovary, oviduct, small intestine, spleen, testis, and uterus. ESTs have been isolated from human B-cell/lung/testis, blood, brain, colon, eye, germ cell, head/neck, heart, heart/melanocyte/uterus, lung, ovary, stomach, testis, and uterus libraries.

Specifications

Antigen LGR6
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6400
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Lgr6
Gene Accession No. Q3UVD5, Q9HBX8
Gene Alias A530037C04Rik; D830026M09; FLJ14471; gonadotropin receptor; GPCR; leucine rich repeat containing G protein-coupled receptor 6; leucine-rich repeat containing G protein-coupled receptor 6; leucine-rich repeat-containing G protein-coupled receptor 6; leucine-rich repeat-containing G-protein coupled receptor 6; Lgr6; RGD1560339; RP11-572A16.3; UNQ6427/PRO21331; VTS20631
Gene Symbols Lgr6
Host Species Rabbit
Immunogen A synthesized peptide derived from human GPCR/LGR6 (850-967AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 329252, 498233, 59352
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less