missing translation for 'onlineSavingsMsg'
Learn More

LGP Consulting Pluggo™ Automated Decapper

Catalog No. 23047100
Change view
Click to view available options
Width (English):
10 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Width (English)
23047100 10 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23047100 Supplier LGP Consulting Supplier No. 4600 4450

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Automated system that safely removes original stoppers from vacuum tubes

  • Removes standard rubber or plastic nonscrew caps: 50 tubes/min. or 3000 tubes/hr.
  • Small benchtop footprint
  • Handles all standard tube sizes, glass or plastic
  • Convenient disposal of caps through adjustable chute

Order Info

Carousels of various sizes available separately (Cat. Nos. 23-047-101 through -103).

Specifications

Width (English) 10 in.
Width (Metric) 25.4 cm
Type Decapper
For Use With (Equipment) Standard size tubes with Rubber or Plastic non-screw caps
For Use With (Application) Ideal to safely remove original stoppers from vacuum tubes