missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Lewis A/B Recombinant Mouse Monoclonal Antibody (HEA164)
GREENER_CHOICE

Catalog No. MA551802
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA551802 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA551802 Supplier Invitrogen™ Supplier No. MA551802
Only null left

Mouse Recombinant Monoclonal Antibody

This antibody is specific for Lewis a,b carbohydrate antigens.

Lewis A and Lewis B are fucose-containing carbohydrates that are expressed on the surface of red blood cells and other tissues in the body. These carbohydrates are part of the larger family of molecules known as Lewis antigens, which play a role in a variety of biological processes, including immune recognition, cell adhesion, and inflammation. The expression of Lewis A and B on red blood cells is determined by the FUT3 gene, which encodes for the fucosyltransferase enzyme that adds the fucose sugar to precursor molecules. In general, individuals with the FUT3 gene mutation that leads to the expression of Lewis A antigen are unable to produce the Lewis B antigen, and vice versa. However, some individuals may be Lewis negative, meaning that they do not express either antigen due to mutations in the FUT2 gene. The presence or absence of Lewis antigens can also have implications for disease susceptibility and treatment, particularly in the context of cancer and other inflammatory conditions, as they can modulate immune responses and promote tumor growth and metastasis.

Specifications

Antigen Lewis A/B
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HEA164
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% ProClin 300
Gene Alias Lewis A; Lewis B
Host Species Mouse
Immunogen This antibody was raised by immunising mice with human carcinoma cells.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgM κ
Show More Show Less