missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Levonorgestrel, 98%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 232.0°C to 239.0°C |
| Loss on Drying | 0.5% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass Bottle |
| Quantity | 1 g |
| Formula Weight | 312.45 |
| Percent Purity | 98% |
| Chemical Name or Material | Levonorgestrel |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Harmful in contact with skin. Harmful if inhaled. Suspected of causing cancer. May damage fertility or the unborn child. |
| Precautionary Statement | GHS P Statement IF ON SKIN: Wash with plenty of soap and water. Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection. |
| EINECSNumber | 212-349-8 |
RUO – Research Use Only