accessibility menu, dialog, popup

UserName

CAS RN 797-63-7

IUPAC Name: (1R,3aS,3bR,9aR,9bS,11aS)-11a-ethyl-1-ethynyl-1-hydroxy-1H,2H,3H,3aH,3bH,4H,5H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
Synonyms: plan b ld norgestrel triagynon norplant II levonorgestrel wyeth brand alcala brand of levonorgestrel microlut preven trinordiol microgyn
Molecular Weight (g/mol): 312.45
Molecular Formula: C21H28O2
InChi Key: WWYNJERNGUHSAO-XUDSTZEESA-N
SMILES: CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1CC[C@@]2(O)C#C
Grade 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results