missing translation for 'onlineSavingsMsg'
Learn More

Levoglucosenone 96.0+%, TCI America™

Supplier:  TCI America L01501G

 View more versions of this product

Catalog No. L01501G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Levoglucosenone
113°C
MFCD00191664
HITOXZPZGPXYHY-UJURSFKZSA-N
(1S,5R)-6,8-dioxabicyclo[3.2.1]oct-2-en-4-one
699486
126.11
Liquid
37112-31-5
C6H6O3
1 g
C1C2C=CC(=O)C(O1)O2
126.111
CHEBI:30999
≥96.0% (GC)

Chemical Identifiers

37112-31-5
126.111
HITOXZPZGPXYHY-UJURSFKZSA-N
CHEBI:30999
C1C2C=CC(=O)C(O1)O2
C6H6O3
MFCD00191664
699486
(1S,5R)-6,8-dioxabicyclo[3.2.1]oct-2-en-4-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : JG7020000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions