missing translation for 'onlineSavingsMsg'
Learn More

Levoglucosenone 96.0+%, TCI America™

Catalog No. p-7131841
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L0150200MG 200 mg
L01501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L0150200MG Supplier TCI America Supplier No. L0150200MG

Chemical Identifiers

CAS 37112-31-5
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.111
MDL Number MFCD00191664
InChI Key HITOXZPZGPXYHY-UJURSFKZSA-N
PubChem CID 699486
ChEBI CHEBI:30999
IUPAC Name (1S,5R)-6,8-dioxabicyclo[3.2.1]oct-2-en-4-one
SMILES C1C2C=CC(=O)C(O1)O2

Specifications

Boiling Point 113°C
Quantity 200 mg
Formula Weight 126.11
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material Levoglucosenone
RTECSNumber JG7020000
TSCA No