missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin USP Hemihydrate, Medisca™

$1389.28 - $5826.99

Chemical Identifiers

CAS 138199-71-0
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
18-048-053
View Documents
Medisca Pharmaceutique Inc
268008
500 g
Each for $5,826.99
Only null left
 
18-048-070
View Documents
Medisca Pharmaceutique Inc
268005
100 g
Each for $1,389.28
Only null left
 
Description

Description

Specifications

Chemical Identifiers

138199-71-0

Specifications

α-Carboxybenzyl Penicillin Disodium Salt
138199-71-0
500 g
USP
Crystalline Powder
White to Light Yellow
98.0 to 102.0%
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'