missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin 98.0+%, TCI America™

Supplier:  TCI America L019325G

 View more versions of this product

Catalog No. L019325G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Levofloxacin
224°C
C18H20FN3O4
25 g
GSDSWSVVBLHKDQ-JTQLQIEISA-N
361.373
CHEBI:63598
≥98.0% (HPLC)
100986-85-4
Yellow
MFCD00865049
levofloxacin, levaquin, quixin, iquix, levofloxacine, cravit, tavanic, --ofloxacin, ofloxacin s---form, s-ofloxacin
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
149096
361.37
Crystalline Powder

Chemical Identifiers

100986-85-4
361.373
GSDSWSVVBLHKDQ-JTQLQIEISA-N
149096
CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O
C18H20FN3O4
MFCD00865049
levofloxacin, levaquin, quixin, iquix, levofloxacine, cravit, tavanic, --ofloxacin, ofloxacin s---form, s-ofloxacin
CHEBI:63598
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UU8815550

TSCA : No

SDS
Documents

Documents

Promotions

Promotions