missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin 98.0+%, TCI America™

Catalog No. p-7131860
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
L01935G 5 g
L019325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. L01935G Supplier TCI America Supplier No. L01935G

Chemical Identifiers

CAS 100986-85-4
Molecular Formula C18H20FN3O4
Molecular Weight (g/mol) 361.373
MDL Number MFCD00865049
InChI Key GSDSWSVVBLHKDQ-JTQLQIEISA-N
Synonym levofloxacin, levaquin, quixin, iquix, levofloxacine, cravit, tavanic, --ofloxacin, ofloxacin s---form, s-ofloxacin
PubChem CID 149096
ChEBI CHEBI:63598
SMILES CC1COC2=C3N1C=C(C(=O)C3=CC(=C2N4CCN(CC4)C)F)C(=O)O

Specifications

Melting Point 224°C
Color Yellow
Quantity 5 g
Formula Weight 361.37
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Levofloxacin
RTECSNumber UU8815550
TSCA No