missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin Related Compound C, Certified Reference Material, MilliporeSigma™ Supelco™

Catalog No. 111023020
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111023020 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111023020 Supplier Merck Emd Millipore Supplier No. PHR223050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control. Provides pharma laboratories and manufacturers with a convenient, and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 177472-30-9
Molecular Formula C20H24FN3O4
Linear Formula C20H24FN3O4
Synonym (3 S)-Ethyl 9-fluoro-2,3-dihydro-3-methyl-10-(4-methyl-1-piperazinyl)-7-oxo-7 H-pyrido[1,2,3-de]-1,4-benzoxazine-6-carboxylate
Quantity 50 mg
Formula Weight 389.42
Grade Certified Reference Material